siRNA Details
| Gene Symbol: Myo10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019472.2 | NP_062345.2 | 39 | 6288 | ||
| GenBank mRNA | AJ249706.2 | - | 39 | 5912 | ||
| AK122371.1 | - | 19 | 4070 | |||
| BC024692.1 | - | 15 | 2414 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Myo10| Gene Symbol: Myo10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_019472.2 | NP_062345.2 | 39 | 6288 | ||
| GenBank mRNA | AJ249706.2 | - | 39 | 5912 | ||
| AK122371.1 | - | 19 | 4070 | |||
| BC024692.1 | - | 15 | 2414 | |||