siRNA Details
| Gene Symbol: Myo7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256081.1 | NP_001243010.1 | 32 | 4450 | ||
| NM_001256082.1 | NP_001243011.1 | 32 | 4297 | |||
| NM_001256083.1 | NP_001243012.1 | 31 | 4279 | |||
| NM_008663.2 | NP_032689.2 | 32 | 4450 | |||
| GenBank mRNA | AY821853.1 | - | 31 | 4286 | ||
| AY821854.1 | - | 32 | 4304 | |||
| U81453.1 | - | 30 | 4200 | |||

Myo7a
Genomic Map