siRNA Details
| Gene Symbol: Myo7a | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256081.1 | NP_001243010.1 | 47 | 6649 | ||
| NM_001256082.1 | NP_001243011.1 | 47 | 6382 | |||
| NM_001256083.1 | NP_001243012.1 | 46 | 6364 | |||
| NM_008663.2 | NP_032689.2 | 47 | 6535 | |||
| GenBank mRNA | AK052607.1 | - | 15 | 2030 | ||
| AY821853.1 | - | 46 | 6371 | |||
| AY821854.1 | - | 47 | 6389 | |||
| U81453.1 | - | 45 | 6399 | |||

Myo7a
Genomic Map