siRNA Details
Gene Symbol: Nab1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008667.3 | NP_032693.2 | 8 | 1667 | ||
GenBank mRNA | AB221677.1 | - | 7 | 1409 | ||
AK018122.1 | - | 2 | 79 | |||
BC005627.1 | - | 8 | 1667 | 3155795 | ||
BC016886.1 | - | 8 | 1590 | 4236565 | ||
U47008.1 | - | 8 | 1545 |
