siRNA Details
| Gene Symbol: Nab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122895.1 | NP_001116367.1 | 4, 5 | 1398 | ||
| NM_008668.2 | NP_032694.2 | 4, 5 | 1398 | |||
| GenBank mRNA | AK158688.1 | - | 4, 5 | 1328 | ||
| AK171180.1 | - | 4, 5 | 1278 | |||
| BC045139.1 | - | 4, 5 | 1282 | 6306757 | ||
| U47543.1 | - | 4, 5 | 1373 | |||

Nab2
Genomic Map