siRNA Details
| Gene Symbol: Nab2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001122895.1 | NP_001116367.1 | 6 | 1595 | ||
| NM_008668.2 | NP_032694.2 | 7 | 1787 | |||
| GenBank mRNA | AK034709.1 | - | 2 | 263 | ||
| AK158688.1 | - | 6 | 1525 | |||
| AK171180.1 | - | 7 | 1667 | |||
| BC045139.1 | - | 7 | 1671 | 6306757 | ||
| U47543.1 | - | 7 | 1762 | |||

Nab2
Genomic Map