siRNA Details
Gene Symbol: Nab2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001122895.1 | NP_001116367.1 | 2 | 533 | ||
NM_008668.2 | NP_032694.2 | 2 | 533 | |||
GenBank mRNA | AK155501.1 | - | 2 | 431 | ||
AK158688.1 | - | 2 | 463 | |||
AK171180.1 | - | 2 | 413 | |||
BC040202.1 | - | 1 | 65 | |||
BC045139.1 | - | 2 | 417 | 6306757 | ||
U47543.1 | - | 2 | 508 |
