siRNA Details
Gene Symbol: Ndufs4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010887.2 | NP_035017.2 | 3 | 279 | ||
GenBank mRNA | AF124785.1 | - | 3 | 229 | ||
AK011124.1 | - | 3 | 267 | |||
AK020552.1 | - | 3 | 235 | |||
BC004618.1 | - | 3 | 237 | 3585543 | ||
CJ041886.1 | - | 3 | 445 | |||
CJ073528.1 | - | 3 | 278 |
