siRNA Details
| Gene Symbol: Ndufs4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010887.2 | NP_035017.2 | 1 | 109 | ||
| GenBank mRNA | AF124785.1 | - | 1 | 59 | ||
| AK011124.1 | - | 1 | 97 | |||
| AK020108.1 | - | 1 | 96 | |||
| AK020552.1 | - | 1 | 65 | |||
| CJ041886.1 | - | 1 | 275 | |||
| CJ073528.1 | - | 1 | 108 | |||

Ndufs4
Genomic Map