siRNA Details
| Gene Symbol: Nedd9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001111324.2 | NP_001104794.1 | 6 | 1438 | ||
| NM_017464.5 | NP_059492.3 | 5 | 1219 | |||
| GenBank mRNA | AF009366.1 | - | 6 | 1374 | ||
| AK030985.1 | - | 6 | 1372 | |||
| AK033729.1 | - | 5 | 1220 | |||
| AK046357.1 | - | 5 | 1182 | |||
| AK054179.1 | - | 5 | 1215 | |||
| AK083374.1 | - | 6 | 1366 | |||
| BC053713.1 | - | 6 | 1371 | 30077589 | ||

Nedd9
Genomic Map