siRNA Details
| Gene Symbol: Nfatc1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001164109.1 | NP_001157581.1 | 2 | 607 | ||
| NM_001164110.1 | NP_001157582.1 | 2 | 607 | |||
| NM_001164111.1 | NP_001157583.1 | 2 | 315 | |||
| NM_001164112.1 | NP_001157584.1 | 2 | 315 | |||
| NM_016791.4 | NP_058071.2 | 2 | 315 | |||
| NM_198429.2 | NP_940821.1 | 2 | 607 | |||
| GenBank mRNA | AF049606.1 | - | 2 | 268 | ||
| AF087434.1 | - | 2 | 302 | |||
| AF239169.1 | - | 2 | 305 | |||
| AK004810.1 | - | 2 | 273 | |||
| AK048610.1 | - | 2 | 609 | |||
| AK144965.1 | - | 2 | 608 | |||
| BB647702.1 | - | 2 | 316 | |||
| BC061509.1 | - | 2 | 594 | 5354603 | ||
| EU887567.1 | - | 2 | 302 | |||
| EU887568.1 | - | 2 | 302 | |||
| EU887569.1 | - | 2 | 260 | |||
| EU887570.1 | - | 2 | 302 | |||
| EU887571.1 | - | 2 | 260 | |||
| EU887572.1 | - | 2 | 260 | |||

Nfatc1
Genomic Map