siRNA Details
| Gene Symbol: Nfkb1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008689.2 | NP_032715.2 | 4 | 557 | ||
| GenBank mRNA | AB119195.1 | - | 2 | 55 | ||
| AB221614.1 | - | 2 | 62 | |||
| AY521462.1 | - | 2 | 55 | |||
| AY521463.1 | - | 2 | 55 | |||
| BC050841.1 | - | 4 | 557 | |||
| BC138535.1 | - | 3 | 134 | 8861555 | ||
| BC138536.1 | - | 3 | 134 | 8861556 | ||
| BC145317.1 | - | 3 | 134 | |||
| M57999.1 | - | 4 | 346 | |||

Nfkb1
Genomic Map