siRNA Details
| Gene Symbol: Nfs1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010911.2 | NP_035041.2 | 12 | 1330 | ||
| GenBank mRNA | AJ222660.1 | - | 12 | 1329 | ||
| AK005724.1 | - | 12 | 1320 | |||
| AK075575.1 | - | 12 | 1328 | |||
| AK132838.1 | - | 12 | 1331 | |||
| AK148527.1 | - | 12 | 1331 | |||
| AK159828.1 | - | 12 | 1319 | |||
| BC010586.1 | - | 12 | 1308 | 3711352 | ||

Nfs1
Genomic Map