siRNA Details
| Gene Symbol: Nid2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008695.2 | NP_032721.2 | 11, 12 | 2815 | ||
| GenBank mRNA | AB017202.1 | - | 11, 12 | 2777 | ||
| AK045306.1 | - | 11, 12 | 2790 | |||
| AK051664.1 | - | 11, 12 | 2791 | |||
| AK052839.1 | - | 11, 12 | 2789 | |||
| AK083629.1 | - | 11, 12 | 2793 | |||
| AK128978.1 | - | 8, 9 | 1937 | |||
| AK140832.1 | - | 11, 12 | 2789 | |||
| AK164258.1 | - | 11, 12 | 2786 | |||
| AY074820.1 | - | 11, 12 | 2776 | |||
| BC054746.1 | - | 11, 12 | 2797 | 6405073 | ||
| BC057016.1 | - | 11, 12 | 2743 | 5719452 | ||

Nid2
Genomic Map