siRNA Details
Gene Symbol: Nkx2-2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001077632.1 | NP_001071100.1 | 2 | 781 | ||
NM_010919.2 | NP_035049.1 | 2 | 781 | |||
GenBank mRNA | AK043561.1 | - | 2 | 635 | ||
AK141002.1 | - | 2 | 786 | |||
BC138159.1 | - | 3 | 602 | 8861178 | ||
BC138160.1 | - | 3 | 602 | 8861179 | ||
U31566.1 | - | 2 | 740 |
