siRNA Details
| Gene Symbol: Nrl | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001136074.2 | NP_001129546.1 | 4 | 838 | ||
| NM_001271916.1 | NP_001258845.1 | 4 | 858 | |||
| NM_001271917.1 | NP_001258846.1 | 3 | 731 | |||
| NM_008736.3 | NP_032762.1 | 3 | 837 | |||
| GenBank mRNA | AK044275.1 | - | 3 | 802 | ||
| AK044316.1 | - | 4 | 821 | |||
| BC031440.1 | - | 4 | 822 | 4527590 | ||
| BC051241.1 | - | 1 | 3869 | |||
| L14935.1 | - | 3 | 776 | |||

Nrl
Genomic Map