siRNA Details
| Gene Symbol: Nrxn2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001205234.1 | NP_001192163.1 | 14 | 3164 | ||
| NM_001205235.1 | NP_001192164.1 | 13 | 3140 | |||
| NM_020253.3 | NP_064649.2 | 14 | 3164 | |||
| GenBank mRNA | AK129239.1 | - | 14 | 3137 | ||
| AK147295.1 | - | 14 | 3100 | |||
| AK147665.1 | - | 14 | 3099 | |||
| BC145496.1 | - | 12 | 2787 | 9054059 | ||
| BC150784.1 | - | 12 | 2787 | 9087695 | ||

Nrxn2
Genomic Map