siRNA Details
| Gene Symbol: Ddr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022563.2 | NP_072075.2 | 15 | 2324 | ||
| GenBank mRNA | AK132504.1 | - | 12 | 1823 | ||
| G49238.1 | - | 16 | 2631 | |||
| X76505.1 | - | 16 | 2631 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Ddr2| Gene Symbol: Ddr2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_022563.2 | NP_072075.2 | 15 | 2324 | ||
| GenBank mRNA | AK132504.1 | - | 12 | 1823 | ||
| G49238.1 | - | 16 | 2631 | |||
| X76505.1 | - | 16 | 2631 | |||