siRNA Details
| Gene Symbol: Odf2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001113213.1 | NP_001106684.1 | 4 | 486 | ||
| NM_001113214.1 | NP_001106685.1 | 4 | 486 | |||
| NM_001177659.1 | NP_001171130.1 | 4 | 486 | |||
| NM_001177661.1 | NP_001171132.1 | 3 | 332 | |||
| NM_013615.3 | NP_038643.1 | 4 | 516 | |||
| GenBank mRNA | AF000968.1 | - | 2 | 306 | ||
| AF034105.1 | - | 4 | 479 | |||
| AK143726.1 | - | 4 | 390 | |||
| AK148523.1 | - | 4 | 399 | |||
| AK165588.1 | - | 4 | 390 | |||
| AK168415.1 | - | 4 | 388 | |||
| BC057001.1 | - | 5 | 543 | 6827260 | ||
| BQ840288.1 | - | 2 | 101 | |||

Odf2
Genomic Map