siRNA Details
| Gene Symbol: Ogn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008760.4 | NP_032786.1 | 5 | 666 | ||
| GenBank mRNA | AK028300.1 | - | 5 | 692 | ||
| AK053650.1 | - | 5 | 667 | |||
| AK075638.1 | - | 4 | 623 | |||
| AK160760.1 | - | 5 | 688 | |||
| AK163772.1 | - | 4 | 618 | |||
| BC021939.1 | - | 4 | 569 | 5067073 | ||
| D31951.1 | - | 4 | 579 | |||

Ogn
Genomic Map