siRNA Details
| Gene Symbol: Ogn | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008760.4 | NP_032786.1 | 5 | 696 | ||
| GenBank mRNA | AK028300.1 | - | 5 | 722 | ||
| AK053650.1 | - | 5 | 697 | |||
| AK075638.1 | - | 4 | 653 | |||
| AK160760.1 | - | 5 | 718 | |||
| AK163772.1 | - | 4 | 648 | |||
| BC021939.1 | - | 4 | 599 | 5067073 | ||
| D31951.1 | - | 4 | 609 | |||

Ogn
Genomic Map