siRNA Details
| Gene Symbol: Olfr19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146335.1 | NP_666447.1 | 1 | 814 | ||
| GenBank mRNA | BC132306.1 | - | 1 | 852 | 40130583 | |
| X89683.1 | - | 1 | 440 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr19| Gene Symbol: Olfr19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146335.1 | NP_666447.1 | 1 | 814 | ||
| GenBank mRNA | BC132306.1 | - | 1 | 852 | 40130583 | |
| X89683.1 | - | 1 | 440 | |||