siRNA Details
| Gene Symbol: Olfr32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010980.2 | NP_035110.1 | 5 | 682 | ||
| GenBank mRNA | BC119404.1 | - | 1 | 457 | 8734153 | |
| BC120653.1 | - | 1 | 457 | 40129576 | ||
| X89678.1 | - | 1 | 34 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr32| Gene Symbol: Olfr32 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010980.2 | NP_035110.1 | 5 | 682 | ||
| GenBank mRNA | BC119404.1 | - | 1 | 457 | 8734153 | |
| BC120653.1 | - | 1 | 457 | 40129576 | ||
| X89678.1 | - | 1 | 34 | |||