siRNA Details
| Gene Symbol: Olfr263 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010984.1 | NP_035114.1 | 1 | 422 | ||
| GenBank mRNA | AF102516.1 | - | 1 | 233 | ||
| BC116918.1 | - | 1 | 501 | 40126237 | ||
| BC120698.1 | - | 1 | 501 | 40129621 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr263| Gene Symbol: Olfr263 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_010984.1 | NP_035114.1 | 1 | 422 | ||
| GenBank mRNA | AF102516.1 | - | 1 | 233 | ||
| BC116918.1 | - | 1 | 501 | 40126237 | ||
| BC120698.1 | - | 1 | 501 | 40129621 | ||