siRNA Details
| Gene Symbol: Olfr47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146370.1 | NP_666482.1 | 1 | 941 | ||
| GenBank mRNA | BC120658.1 | - | 1 | 973 | 40129581 | |
| BC120660.1 | - | 1 | 973 | 40129583 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr47| Gene Symbol: Olfr47 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146370.1 | NP_666482.1 | 1 | 941 | ||
| GenBank mRNA | BC120658.1 | - | 1 | 973 | 40129581 | |
| BC120660.1 | - | 1 | 973 | 40129583 | ||