siRNA Details
| Gene Symbol: Olfr51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146909.1 | NP_667120.1 | 1 | 815 | ||
| GenBank mRNA | AF102525.1 | - | 1 | 626 | ||
| BC111814.1 | - | 1 | 814 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Olfr51| Gene Symbol: Olfr51 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_146909.1 | NP_667120.1 | 1 | 815 | ||
| GenBank mRNA | AF102525.1 | - | 1 | 626 | ||
| BC111814.1 | - | 1 | 814 | |||