siRNA Details
| Gene Symbol: Orm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011016.2 | NP_035146.1 | 1 | 50 | ||
| GenBank mRNA | BC057985.1 | - | 1 | 25 | 5126236 | |
| CB949119.1 | - | 1 | 53 | |||
| M27009.1 | - | 1 | 15 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Orm2| Gene Symbol: Orm2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011016.2 | NP_035146.1 | 1 | 50 | ||
| GenBank mRNA | BC057985.1 | - | 1 | 25 | 5126236 | |
| CB949119.1 | - | 1 | 53 | |||
| M27009.1 | - | 1 | 15 | |||