siRNA Details
| Gene Symbol: Hspa4l | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011020.3 | NP_035150.3 | 8 | 1016 | ||
| GenBank mRNA | AB001926.1 | - | 8 | 982 | ||
| AK144225.1 | - | 8 | 1053 | |||
| AK163459.1 | - | 9 | 1188 | |||
| BC012712.1 | - | 8 | 1085 | 4190161 | ||
| BC057002.1 | - | 8 | 1016 | 5702602 | ||
| BC110662.1 | - | 8 | 1036 | 4511154 | ||
| D49482.1 | - | 8 | 1010 | |||
| U23921.1 | - | 8 | 1133 | |||

Hspa4l
Genomic Map