siRNA Details
| Gene Symbol: Pabpc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011033.2 | NP_035163.1 | 1 | 1847 | ||
| GenBank mRNA | BC051134.1 | - | 1 | 1827 | 6704229 | |
| X75959.1 | - | 1 | 1823 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pabpc2| Gene Symbol: Pabpc2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_011033.2 | NP_035163.1 | 1 | 1847 | ||
| GenBank mRNA | BC051134.1 | - | 1 | 1827 | 6704229 | |
| X75959.1 | - | 1 | 1823 | |||