siRNA Details
| Gene Symbol: Pafah1b1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_013625.4 | NP_038653.1 | 5 | 886 | ||
| GenBank mRNA | AK144908.1 | - | 5 | 875 | ||
| AK147207.1 | - | 5 | 824 | |||
| AK147229.1 | - | 5 | 837 | |||
| AK147237.1 | - | 5 | 878 | |||
| AK147309.1 | - | 5 | 821 | |||
| AK147507.1 | - | 5 | 1160 | |||
| AK147515.1 | - | 5 | 873 | |||
| AK147533.1 | - | 5 | 853 | |||
| AK147793.1 | - | 5 | 548 | |||
| AK164008.1 | - | 5 | 557 | |||
| AK167448.1 | - | 5 | 853 | |||
| AK167824.1 | - | 5 | 878 | |||
| AK168929.1 | - | 5 | 526 | |||
| AK168960.1 | - | 5 | 841 | |||
| AY189217.1 | - | 4 | 505 | |||
| AY189218.1 | - | 4 | 505 | |||
| BC014831.1 | - | 5 | 825 | 4507474 | ||
| BC026141.1 | - | 5 | 850 | 4017963 | ||
| BC046989.1 | - | 5 | 858 | |||
| BC099462.1 | - | 5 | 824 | |||
| L25108.1 | - | 5 | 786 | |||
| U95116.1 | - | 5 | 1181 | |||

Pafah1b1
Genomic Map