siRNA Details
| Gene Symbol: Pafah1b2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008775.3 | NP_032801.2 | 2, 3 | 266 | ||
| GenBank mRNA | AK012116.1 | - | 2, 3 | 180 | ||
| AK012164.1 | - | 2, 3 | 201 | |||
| AK153424.1 | - | 2, 3 | 237 | |||
| BB856076.1 | - | 2, 3 | 267 | |||
| BC002037.1 | - | 2, 3 | 214 | 3486003 | ||
| BC056211.2 | - | 2, 3 | 236 | 30012448 | ||
| U57747.1 | - | 2, 3 | 198 | |||

Pafah1b2
Genomic Map