siRNA Details
Gene Symbol: Pafah1b2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_008775.3 | NP_032801.2 | 6 | 607 | ||
GenBank mRNA | AK153424.1 | - | 6 | 578 | ||
BC002037.1 | - | 6 | 555 | 3486003 | ||
BC056211.2 | - | 6 | 577 | 30012448 | ||
U57747.1 | - | 6 | 539 |
