siRNA Details
| Gene Symbol: Pax5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008782.2 | NP_032808.1 | 9 | 1083 | ||
| GenBank mRNA | M97013.1 | - | 9 | 1083 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pax5| Gene Symbol: Pax5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008782.2 | NP_032808.1 | 9 | 1083 | ||
| GenBank mRNA | M97013.1 | - | 9 | 1083 | ||