siRNA Details
| Gene Symbol: Pcbp2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001103165.1 | NP_001096635.1 | 13, 14 | 1129 | ||
| NM_001103166.1 | NP_001096636.1 | 12, 13 | 1036 | |||
| NM_001174073.1 | NP_001167544.1 | 11, 12 | 985 | |||
| NM_011042.2 | NP_035172.2 | 12, 13 | 1090 | |||
| GenBank mRNA | AK006450.1 | - | 2, 3 | 95 | ||
| AK012297.1 | - | 13, 14 | 976 | |||
| AK161527.1 | - | 11, 12 | 833 | |||
| BC020478.1 | - | 11, 12 | 781 | |||
| EU604548.1 | - | 12, 13 | 881 | |||
| L19661.1 | - | 13, 14 | 1016 | |||

Pcbp2
Genomic Map