siRNA Details
| Gene Symbol: Pcnt | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001282992.1 | NP_001269921.1 | 23 | 4445 | ||
| NM_008787.3 | NP_032813.3 | 23 | 4445 | |||
| GenBank mRNA | AB207233.1 | - | 23 | 4319 | ||
| AB207234.1 | - | 11 | 1740 | |||
| AK122275.2 | - | 8 | 1192 | |||

Pcnt
Genomic Map