siRNA Details
| Gene Symbol: Pcsk5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001163144.1 | NP_001156616.1 | 19 | 2942 | ||
| NM_001190483.1 | NP_001177412.1 | 19 | 2942 | |||
| GenBank mRNA | AK143998.1 | - | 18 | 2839 | ||
| BC013068.1 | - | 19 | 2562 | |||
| D12619.1 | - | 19 | 2531 | |||
| D17583.1 | - | 12 | 1475 | |||
| L14932.1 | - | 19 | 2492 | |||

Pcsk5
Genomic Map