siRNA Details
| Gene Symbol: Pcsk5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190483.1 | NP_001177412.1 | 31, 32 | 4592 | ||
| GenBank mRNA | D17583.1 | - | 24, 25 | 3125 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pcsk5| Gene Symbol: Pcsk5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001190483.1 | NP_001177412.1 | 31, 32 | 4592 | ||
| GenBank mRNA | D17583.1 | - | 24, 25 | 3125 | ||