siRNA Details
| Gene Symbol: Pcsk7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001281934.1 | NP_001268863.1 | 16 | 2835 | ||
| NM_008794.2 | NP_032820.2 | 16 | 2571 | |||
| GenBank mRNA | AK148416.1 | - | 16 | 2156 | ||
| AK151980.1 | - | 16 | 2158 | |||
| AK165389.1 | - | 16 | 2177 | |||
| AK177684.1 | - | 1 | 92 | |||
| AK207794.1 | - | 1 | 55 | |||
| BC006730.1 | - | 15 | 2072 | 3962441 | ||
| BC007322.1 | - | 16 | 2432 | |||
| BC056456.1 | - | 17 | 2645 | |||
| U48830.1 | - | 16 | 2143 | |||

Pcsk7
Genomic Map