siRNA Details
| Gene Symbol: Pcx | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001162946.1 | NP_001156418.1 | 14 | 1928 | ||
| NM_008797.3 | NP_032823.2 | 13 | 1905 | |||
| GenBank mRNA | AK077665.1 | - | 5 | 733 | ||
| AK128927.1 | - | 1855 | ||||
| AK148328.1 | - | 14 | 1875 | |||
| AK170596.1 | - | 14 | 1888 | |||
| AK170871.1 | - | 14 | 1878 | |||
| AK172320.1 | - | 14 | 1856 | |||
| BC055030.1 | - | 14 | 1863 | 5053475 | ||
| L09192.1 | - | 13 | 1881 | |||
| M97957.1 | - | 8 | 1053 | |||

Pcx
Genomic Map