siRNA Details
| Gene Symbol: Pdcd4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168491.1 | NP_001161963.1 | 6, 7 | 981 | ||
| NM_001168492.1 | NP_001161964.1 | 6, 7 | 981 | |||
| NM_011050.4 | NP_035180.2 | 6, 7 | 1069 | |||
| GenBank mRNA | AK134366.1 | - | 6, 7 | 1070 | ||
| AK172654.1 | - | 6, 7 | 937 | |||
| BC055739.1 | - | 6, 7 | 916 | 6414089 | ||
| CT010188.1 | - | 5, 6 | 779 | |||
| D50465.1 | - | 5, 6 | 849 | |||
| D86344.1 | - | 6, 7 | 865 | |||

Pdcd4
Genomic Map