siRNA Details
| Gene Symbol: Pde6d | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008801.2 | NP_032827.1 | 4, 5 | 520 | ||
| GenBank mRNA | AF039216.1 | - | 4, 5 | 479 | ||
| AK141209.1 | - | 4, 5 | 526 | |||
| AK165701.1 | - | 4, 5 | 745 | |||
| AK170067.1 | - | 4, 5 | 532 | |||
| BC005636.1 | - | 4, 5 | 520 | 3964336 | ||

Pde6d
Genomic Map