siRNA Details
| Gene Symbol: Pde6b | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008806.2 | NP_032832.2 | 1 | 343 | ||
| GenBank mRNA | AK039397.1 | - | 1 | 55 | ||
| AK044364.1 | - | 1 | 286 | |||
| BC048247.1 | - | 1 | 322 | |||
| BC058803.1 | - | 1 | 290 | |||
| BC129923.1 | - | 1 | 273 | 40111882 | ||
| BC129924.1 | - | 1 | 272 | |||
| BU505563.1 | - | 1 | 343 | |||
| X55968.1 | - | 1 | 276 | |||
| X60133.1 | - | 1 | 283 | |||
| X87952.1 | - | 1 | 283 | |||

Pde6b
Genomic Map