siRNA Details
| Gene Symbol: Per3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289877.1 | NP_001276806.1 | 17 | 3168 | ||
| NM_001289878.1 | NP_001276807.1 | 18 | 3217 | |||
| NM_011067.3 | NP_035197.2 | 17 | 3165 | |||
| GenBank mRNA | AB013605.1 | - | 18 | 2972 | ||
| AF050182.1 | - | 17 | 3089 | |||
| AJ007377.1 | - | 18 | 2972 | |||
| AK084610.1 | - | 9 | 4014 | |||
| AK139878.1 | - | 3 | 966 | |||
| AK166514.1 | - | 18 | 3396 | |||
| AK173261.1 | - | 5 | 4455 | |||
| BC141032.1 | - | 18 | 2766 | 9055644 | ||
| BC145178.1 | - | 19 | 2815 | |||

Per3
Genomic Map