siRNA Details
| Gene Symbol: Per3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001289877.1 | NP_001276806.1 | 19 | 3395 | ||
| NM_001289878.1 | NP_001276807.1 | 20 | 3444 | |||
| NM_011067.3 | NP_035197.2 | 19 | 3392 | |||
| GenBank mRNA | AB013605.1 | - | 20 | 3205 | ||
| AF050182.1 | - | 19 | 3316 | |||
| AJ007377.1 | - | 20 | 3205 | |||
| AK139878.1 | - | 5 | 1193 | |||
| BC141032.1 | - | 20 | 2999 | 9055644 | ||
| BC145178.1 | - | 21 | 3048 | |||

Per3
Genomic Map