siRNA Details
| Gene Symbol: Pgr | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008829.2 | NP_032855.2 | 1 | 1751 | ||
| GenBank mRNA | AK036862.1 | - | 1 | 1166 | ||
| AK054117.1 | - | 1 | 432 | |||
| AK083181.1 | - | 1 | 233 | |||
| BC145807.1 | - | 1 | 1156 | 40131110 | ||
| M68915.1 | - | 1 | 1117 | |||

Pgr
Genomic Map