siRNA Details
| Gene Symbol: Abcb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008830.2 | NP_032856.2 | 21 | 2674 | ||
| GenBank mRNA | AK050345.1 | - | 20 | 2627 | ||
| BC150687.1 | - | 21 | 2544 | 9087598 | ||
| CJ071650.1 | - | 3 | 225 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Abcb4| Gene Symbol: Abcb4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008830.2 | NP_032856.2 | 21 | 2674 | ||
| GenBank mRNA | AK050345.1 | - | 20 | 2627 | ||
| BC150687.1 | - | 21 | 2544 | 9087598 | ||
| CJ071650.1 | - | 3 | 225 | |||