siRNA Details
| Gene Symbol: Pik3ca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008839.2 | NP_032865.2 | 8 | 1485 | ||
| GenBank mRNA | AK051885.1 | - | 8 | 1118 | ||
| BC089038.1 | - | 8 | 1485 | 30692343 | ||
| BC130228.1 | - | 8 | 1434 | 40141870 | ||
| U03279.1 | - | 8 | 1430 | |||

Pik3ca
Genomic Map