siRNA Details
| Gene Symbol: Pim2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138606.2 | NP_613072.1 | 5 | 817 | ||
| GenBank mRNA | BC022727.1 | - | 2 | 358 | ||
| BC027376.1 | - | 5 | 737 | 4954711 | ||
| L41495.1 | - | 5 | 817 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pim2| Gene Symbol: Pim2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138606.2 | NP_613072.1 | 5 | 817 | ||
| GenBank mRNA | BC022727.1 | - | 2 | 358 | ||
| BC027376.1 | - | 5 | 737 | 4954711 | ||
| L41495.1 | - | 5 | 817 | |||