siRNA Details
| Gene Symbol: Prkaca | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001277898.1 | NP_001264827.1 | 9, 10 | 1100 | ||
| NM_008854.5 | NP_032880.1 | 9, 10 | 1143 | |||
| GenBank mRNA | AK188175.1 | - | 1, 2 | 124 | ||
| BC003238.1 | - | 9, 10 | 1133 | 3497908 | ||
| BC054834.1 | - | 9, 10 | 1128 | 6439034 | ||
| CB248596.1 | - | 1, 2 | 40 | |||
| CD807077.1 | - | 5, 6 | 547 | |||
| M12303.1 | - | 9, 10 | 1120 | |||

Prkaca
Genomic Map