siRNA Details
| Gene Symbol: Pla2g2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008868.3 | NP_032894.2 | 4 | 462 | ||
| GenBank mRNA | BC029347.1 | - | 3 | 307 | 4234596 | |
| CA464794.1 | - | 4 | 465 | |||
| U18119.1 | - | 3 | 288 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

Pla2g2c| Gene Symbol: Pla2g2c | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_008868.3 | NP_032894.2 | 4 | 462 | ||
| GenBank mRNA | BC029347.1 | - | 3 | 307 | 4234596 | |
| CA464794.1 | - | 4 | 465 | |||
| U18119.1 | - | 3 | 288 | |||